Learn More
Medchemexpress LLC Insulin β Chain Peptide (15-23) | 247044-67-3 | 95.7% | 1009.18 | 5 MG
SDP
Catalog No. 5000424245
Encompass_Preferred
Insulin β Chain Peptide (15-23) is one of the earliest antigenic epitopes to which CD8 T-cells respond. This peptide is for research use only and not intended for patient use. It is a white to off-white solid with a purity of 95.68%.
  • Exhibits high purity at 95.68%
  • For research use only
  • Stimulates IFN-γ and TNF-α production in CD8 T-cells in vitro
  • Promotes rapid diabetes development in specific mouse models in vivo
  • White to off-white solid appearance
  • Features amino acid sequence Leu-Tyr-Leu-Val-Cys-Gly-Glu-Arg-Gly (LYLVCGERG)
Catalog No. 50-004-24245 Supplier Medchemexpress LLC Supplier No. HYP25115MG
Insulin β Chain Peptide (15-23) is one of the earliest antigenic epitopes to which CD8 T-cells respond. This peptide is for research use only and not intended for patient use. It is a white to off-white solid with a purity of 95.68%.
  • Exhibits high purity at 95.68%
  • For research use only
  • Stimulates IFN-γ and TNF-α production in CD8 T-cells in vitro
  • Promotes rapid diabetes development in specific mouse models in vivo
  • White to off-white solid appearance
  • Features amino acid sequence Leu-Tyr-Leu-Val-Cys-Gly-Glu-Arg-Gly (LYLVCGERG)