Learn More
Alpco Ltd Insulin Chemiluminescence ELISA Jumbo Pack (10 Plates)
SDP
Supplier:  Alpco Ltd 80INSHUCH10
Encompass
The ALPCO Chemi Human Insulin ELISA Jumbo Pack (10 Plates) measures the concentration of insulin in human serum, heparin plasma, and tissue culture supernatants.
  • Broad dynamic range of 5–30,000 pg/mL
  • Superior sensitivity compared other products on the market. 
  • LOD: <2.0 pg/mL
  • LLOQ: 20 pg/mL
FDA Class I Device. For In Vitro Diagnostic Use.
Catalog No. NC2271694