Learn More
Kamiya Biomedical Insulin Control, L1: 2 x 0.5mL, L2: 2 x 0.5mL RX
SDP
Catalog No. NC1682365 Shop All Kamiya Biomedical Products
Encompass_Preferred
Insulin Control; L1 & L2: 2 x 0.5mL each, lyophilized. The K-ASSAY Insulin Control is intended for use as consistent test sample of known concentration for monitoring the performance of the K-ASSAY Insulin immunoturbidimetric assay (Cat. No. KAI-071). FOR IN VITRO DIAGNOSTIC USE.
This item is not returnable. View return policy
Catalog No. NC1682365 Supplier Kamiya Biomedical Supplier No. K73C4M
 Restricted Item
Add to Cart
Add to Cart
Insulin Control; L1 & L2: 2 x 0.5mL each, lyophilized. The K-ASSAY Insulin Control is intended for use as consistent test sample of known concentration for monitoring the performance of the K-ASSAY Insulin immunoturbidimetric assay (Cat. No. KAI-071). FOR IN VITRO DIAGNOSTIC USE.