Learn More
Drg International Inc DRG Insulin ELISA, 96 Well Kit
Supplier:  Drg International Inc EIA2935
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of insulin in serum and plasma. Insulin is the principal hormone responsible for the control of glucose metabolism. The DRG Insulin ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal antibody directed towards a unique antigenic site on the Insulin molecule.
Catalog No. 50-187-1255