Learn More
Enzo Life Sciences Insulin ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ENZKIT1410001
Encompass_Preferred
The Insulin ELISA kit is a colorimetric immunometric immunoassay kit with results in 3 hours.  Assay Time: 3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human Porcine.
Catalog No. 50-201-3835