Learn More
Crystal Chem Inc Insulin ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 90095
Encompass
The Insulin ELISA is a highly sensitive assay used to quantify levels of human insulin in serum and plasma.Highlights: ~100% Cross-reactivity to insulin and most analogs. Very low (1.2%) reactivity to proinsulin. Very low (< 5%) reactivity to mouse/rat insulin. Fast assay time of 2 hrs 15 mins. Wide dyanamic range of 0.9 - 220 mU/L. For bulk purchases save money with our 10-kit pack.Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7922