Learn More
Cygnus Technologies Insulin ELISA Kit, Ultra-Sensitive
Supplier:  Cygnus Technologies F280
Encompass
This kit is intended for use in quantitating very low concentrations of insulin, in the range of 25 to 1250 pg/ml, from bovine, human, or porcine sources.
Catalog No. NC1730170