Learn More
Waters Corp Waters Insulin Diol (OH) Column, 125Å, 10 μm, 7.8 mm, 300 mm
Supplier: Waters Corp WAT201549
This insulin diol (OH) column is for manufacturing and quality control tests on insulin products. It analyzes impurities with molecular masses greater than insulin, offering faster run times, higher sensitivity, and improved resolution of insulin and covalent insulin HMW, while reducing acetonitrile-containing waste stream volumes.
- Optimized for insulin product manufacturing and quality control.
- Analyzes impurities with molecular masses greater than insulin.
- Features faster run times, higher sensitivity, and improved resolution for insulin and HMW.
- Reduces acetonitrile-containing waste stream volumes.
- Particle size: 10 μm; Pore size: 125 Å.
- Dimensions: 7.8 mm inner diameter x 300 mm length.
- Max pressure: 6000 psi (415 bar); pH range: 2 to 8.
- Molecular weight range: 5000 to 150000.
- Contains hazardous material.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.