Learn More
Medchemexpress LLC Insulin (human) | 11061-68-0 | 99.9% | 5807.57 | 200 MG
SDP
Catalog No. 5000425429
Encompass_Preferred
Insulin (human) is a polypeptide hormone vital for regulating glucose levels in the body, primarily used in the treatment of diabetes mellitus. This product is a recombinant human insulin, a synthetic version of the hormone, and appears as a white to off-white solid. The human insulin gene contains specific intervening sequences, one within the 5'-untranslated segment of the mRNA and another interrupting the C-peptide encoding region, which are important for its biological function.
  • Regulates glucose levels
  • Used for diabetes mellitus treatment
  • Recombinant human insulin
  • White to off-white solid appearance
Catalog No. 50-004-25429 Supplier Medchemexpress LLC Supplier No. HYP0035200MG
Insulin (human) is a polypeptide hormone vital for regulating glucose levels in the body, primarily used in the treatment of diabetes mellitus. This product is a recombinant human insulin, a synthetic version of the hormone, and appears as a white to off-white solid. The human insulin gene contains specific intervening sequences, one within the 5'-untranslated segment of the mRNA and another interrupting the C-peptide encoding region, which are important for its biological function.
  • Regulates glucose levels
  • Used for diabetes mellitus treatment
  • Recombinant human insulin
  • White to off-white solid appearance