Learn More
TARGETMOL CHEMICALS INC Insulin human 200MG
SDP
Catalog No. 502876669
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Insulin (human) is a peptide hormone that promotes glycogen synthesis and regulates glucose levels in the blood. Insulin (human) has hypoglycemic activity and is used clinically to treat hyperglycemia in diabetic patients. Purity 99.99%
Catalog No. 50-287-6669 Supplier TARGETMOL CHEMICALS INC Supplier No. T8221200MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Insulin (human) is a peptide hormone that promotes glycogen synthesis and regulates glucose levels in the blood. Insulin (human) has hypoglycemic activity and is used clinically to treat hyperglycemia in diabetic patients. Purity 99.99%