Learn More
BioVendor, LLC Insulin Human ELISA (96 wells)
SDP
Supplier:  BioVendor, LLC RIS006R
Encompass
Insulin Human ELISA, 96 wells. Research use only. Immunoassay kit to quantify human insulin in serum. Cross-reactivity with some animal insulins (see manual for specifics). Quality controls included. Calibration range: 5.1250 uIU/ml. TypeSandwich ELISA, HRP-labelled antibodyApplicationsSerumSample Requirements50 µl/wellCalibration Range5.1–250 µIU/mlLimit of Detection0.17 µIU/mlSummary: Insulin is a peptide hormone secreted from B cells of islet of Langerhans in the pancreas. Main targets of insulin are liver, muscle, and adipose tissue. In the liver, it promotes glycogenesis, protein synthesis, fatty acid synthesis, carbohydrate utilization, and inhibition of gluconeogenesis. In the muscle, it promotes membrane permeability for carbohydrates, amino acids and K ion, glycogenesis, protein synthesis, while inhibits protein degradation. In the adipose tissue, it promotes membrane permeability for glucose and fatty acid synthesis.
Catalog No. 50-206-9260