Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Insulin, Human Recombinant

Catalog No. MP219390080 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219390080 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219390080 Supplier MP Biomedicals™ Supplier No. 0219390080
Add to Cart
Add to Cart

Insulin, Human Recombinan, ∽28 IU/mg, Powder

  • Methods for immobilizing insulin on polystyrene dishes have been reported for applications involving protein-free cell cultures.
  • Insulin is the primary polypeptide hormone responsible for controlling the cellular uptake, utilization and storage of glucose, amino acids and fatty acids while inhibiting the breakdown of glycogen, protein and fat.
  • It is produced by pancreatic beta cells.
TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 5807.6
Quantity 100 mg
Storage Requirements Store at -20°C.
Product Type Proteins, Enzymes & Peptides
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.