Learn More
Sino Biological Insulin / INS Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11038MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Insulin / INS (rh Insulin / INS; Catalog#11038-HNAY; P01308; Phe1-Lys29 & Gly1-Asn21). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8164