Learn More
Alpco Ltd Insulin ELISA Jumbo
SDP
Supplier:  Alpco Ltd 80INSHUE10.1
Encompass
The Insulin ELISA Jumbo Pack (10 plates) quantifies the concentration of insulin protein in human serum or plasma samples. FDA Class I Device. For In Vitro Diagnostic Use.
  • Only 25 µL of sample is sufficient for the assay
  • Superior sensitivity of 0.399 µIU/mL
  • Dynamic range of 3.0-200 µIU/mL
Catalog No. 50-751-3654