Learn More
Cusabio Technology LLC insulin-like 3 Leydig cell
Catalog No. 5000048899
Encompass_Preferred
This kit detects INSL3 (insulin-like 3) in human serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 0 312 ng/mL to 20 ng/mL with a sensitivity of 0 078 ng/mL enabling precise measurement of low-abundance targets The assay requires 50-100 L of sample per test and delivers results within 1-5 hours Optical density is measured at 450 nm to determine INSL3 concentrations providing researchers with a reliable tool for studying reproductive biology endocrine disorders and developmental processes The kit s standardized protocol ensures consistent performance across diverse sample types while maintaining high specificity for the target analyte
Catalog No. 50-000-48899 Supplier Cusabio Technology LLC Supplier No. CSBE13909H
This kit detects INSL3 (insulin-like 3) in human serum plasma and tissue homogenates using a quantitative sandwich ELISA method It offers a detection range of 0 312 ng/mL to 20 ng/mL with a sensitivity of 0 078 ng/mL enabling precise measurement of low-abundance targets The assay requires 50-100 L of sample per test and delivers results within 1-5 hours Optical density is measured at 450 nm to determine INSL3 concentrations providing researchers with a reliable tool for studying reproductive biology endocrine disorders and developmental processes The kit s standardized protocol ensures consistent performance across diverse sample types while maintaining high specificity for the target analyte