Learn More
Cusabio Technology LLC Insulin-like Growth Factor 1
Catalog No. 5000051906
Encompass_Preferred
This ELISA kit is designed for the quantitative detection of Monkey Insulin-like Growth Factor 1 (IGF-1) in serum and plasma samples using a sandwich assay method It offers a broad detection range of 62 5 ng/mL to 4000 ng/mL with a sensitivity of 15 6 ng/mL ensuring precise measurement of physiological and pathological IGF-1 levels The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours making it suitable for both routine analysis and time-sensitive research applications Measurements are performed at an absorbance wavelength of 450 nm providing reliable and reproducible results for studies involving growth regulation metabolic processes or disease mechanisms in primate models
Catalog No. 50-000-51906 Supplier Cusabio Technology LLC Supplier No. CSBE13401MK
This ELISA kit is designed for the quantitative detection of Monkey Insulin-like Growth Factor 1 (IGF-1) in serum and plasma samples using a sandwich assay method It offers a broad detection range of 62 5 ng/mL to 4000 ng/mL with a sensitivity of 15 6 ng/mL ensuring precise measurement of physiological and pathological IGF-1 levels The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours making it suitable for both routine analysis and time-sensitive research applications Measurements are performed at an absorbance wavelength of 450 nm providing reliable and reproducible results for studies involving growth regulation metabolic processes or disease mechanisms in primate models