Learn More
Cusabio Technology LLC Insulin-like growth factor 2
Catalog No. 5000048218
Encompass_Preferred
This kit detects IGF2 in serum plasma and tissue homogenates with a quantitative sandwich ELISA methodology offering a detection range of 78 pg/mL to 5000 pg/mL and a sensitivity of 19 5 pg/mL The assay requires only 50-100 L of sample per test and delivers results within 1 to 5 hours Optimized for absorbance measurements at 450 nm it provides reliable and precise quantification of IGF2 levels in diverse biological matrices Designed for research applications this kit ensures efficient performance while maintaining accuracy across its broad analytical range making it suitable for studies requiring robust measurement of IGF2 concentrations in complex samples
Catalog No. 50-000-48218 Supplier Cusabio Technology LLC Supplier No. CSBE04585R
This kit detects IGF2 in serum plasma and tissue homogenates with a quantitative sandwich ELISA methodology offering a detection range of 78 pg/mL to 5000 pg/mL and a sensitivity of 19 5 pg/mL The assay requires only 50-100 L of sample per test and delivers results within 1 to 5 hours Optimized for absorbance measurements at 450 nm it provides reliable and precise quantification of IGF2 levels in diverse biological matrices Designed for research applications this kit ensures efficient performance while maintaining accuracy across its broad analytical range making it suitable for studies requiring robust measurement of IGF2 concentrations in complex samples