Learn More
Biomatik Corporation Insulin Like Growth Factor Binding Protein 4 (Igfbp4) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29153
Encompass_Preferred
Insulin Like Growth Factor Binding Protein 4 (Igfbp4) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Insulin Like Growth Factor Binding Protein 4 (IGFBP4). The Insulin Like Growth Factor Binding Protein 4 (Igfbp4) Monoclonal Antibody has been validated for the following applications: WB. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Asp22~Glu258. Applications: WB. Target Synonyms: Insulin Like Growth Factor Binding Protein 4 (IGFBP4). Accession Number: P22692. Gene ID: 3487. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9217