Learn More
Cusabio Technology LLC Insulin-like growth factor II
Catalog No. 5000051788
Encompass_Preferred
This kit detects IGF2 in serum and plasma samples with a detection range of 62 5 pg/mL to 1000 pg/mL providing reliable quantitative results through a competitive ELISA method It offers a sensitivity of 31 25 pg/mL ensuring accurate measurement of low-concentration samples The assay requires a sample volume of 50-100 L and can be completed within 1 to 5 hours depending on protocol optimization Results are determined spectrophotometrically at 450 nm making it compatible with standard laboratory equipment Designed for research applications this kit is suitable for studying insulin-like growth factor II dynamics in fish models supporting studies in growth regulation metabolism and developmental biology
Catalog No. 50-000-51788 Supplier Cusabio Technology LLC Supplier No. CSBEL011088FI
This kit detects IGF2 in serum and plasma samples with a detection range of 62 5 pg/mL to 1000 pg/mL providing reliable quantitative results through a competitive ELISA method It offers a sensitivity of 31 25 pg/mL ensuring accurate measurement of low-concentration samples The assay requires a sample volume of 50-100 L and can be completed within 1 to 5 hours depending on protocol optimization Results are determined spectrophotometrically at 450 nm making it compatible with standard laboratory equipment Designed for research applications this kit is suitable for studying insulin-like growth factor II dynamics in fish models supporting studies in growth regulation metabolism and developmental biology