Learn More
Alpco Ltd Mouse Insulin ELISA
SDP
Supplier:  Alpco Ltd 80INSMSE01
Encompass_Preferred
The Mouse Insulin ELISA quantifies the concentration of insulin protein products in mouse I and mouse II proinsulin genes with just 10 µL of serum or plasma sample. For Research Use Only. Not for Use in Diagnostic Procedures.
  • Low sample volume of 10 µL
  • Dynamic Range of 0.188-6.9 ng/mL
  • Superior sensitivity of 0.06 ng/mL
Catalog No. 50-922-584