Learn More
Alpco Ltd Mouse Ultrasensitive Insulin ELISA
SDP
Supplier:  Alpco Ltd 80INSMSUE01
Encompass_Preferred
The Mouse Ultrasensitive Insulin ELISA quantifies the concentration of insulin protein products in mouse I and mouse II proinsulin genes from as little as 5 µL of serum or plasma sample. For Research Use Only. Not for Use in Diagnostic Procedures.
  • Only 5 µL of sample required for tail snips
  • Dynamic range of 0.18-6.9 ng/mL
  • Superior sensitivity of 0.115 ng/mL
  • Only 25 µL of sample required for hypoinsulinemic samples
  • Dynamic range of 0.025-1.25 ng/mL
  • Superior sensitivity of 0.019 ng/mL
Catalog No. 50-922-588