Learn More
Carbosynth LLC. Purified recombinant Human Insulin protein
SDP
Supplier:  Carbosynth LLC. 30AI5125MG
Encompass_Preferred
Insulin is a hormone that is central to regulating carbohydrate and fat metabolism in the body. Insulin causes cells in the liver, muscle, and fat tissue to take up glucose from the blood, storing it as glycogen in the liver and muscles.
Catalog No. 50-125-3048