Learn More
Medchemexpress LLC Insulin R/CD220 Hum 5ug
SDP
Catalog No. 5000179782
Encompass_Preferred
The insulin R/CD220 protein is a receptor tyrosine kinase that coordinates insulin action by phosphorylating substrates such as IRS and SHC upon insulin binding Phosphorylated IRS activates PI3K-AKT/PKB to exert metabolic effects and activates Ras-MAPK to promote growth and differentiation Insulin R/CD220 Protein Human (HEK293 C-His) is the recombinant human-derived Insulin R/CD220 protein expressed by HEK293 with C-10 His labeled tag
Catalog No. 50-001-79782 Supplier Medchemexpress LLC Supplier No. HYP73873A5UG
The insulin R/CD220 protein is a receptor tyrosine kinase that coordinates insulin action by phosphorylating substrates such as IRS and SHC upon insulin binding Phosphorylated IRS activates PI3K-AKT/PKB to exert metabolic effects and activates Ras-MAPK to promote growth and differentiation Insulin R/CD220 Protein Human (HEK293 C-His) is the recombinant human-derived Insulin R/CD220 protein expressed by HEK293 with C-10 His labeled tag