Learn More
Alpco Ltd Rat Insulin ELISA
SDP
Supplier:  Alpco Ltd 80INSRTE01
Encompass_Preferred
The Rat Insulin ELISA is for the quantitative determination of insulin in rat serum and plasma. For Research Use Only. Not for Use in Diagnostic Procedures.
  • Low sample volume of 10 µL
  • Dynamic Range of 0.15-5.5 ng/mL
  • Superior sensitivity of 0.124 ng/mL
Catalog No. 50-922-592