Learn More
Mercodia Inc INSULIN RAT ELISA
SDP
Supplier:  Mercodia Inc 10125001
Encompass_Preferred
Mercodia Rat Insulin ELISA provides a method for the quantitative determination of insulin in mouse serum, plasma, or cell culture media. The Mercodia Rat Insulin ELISA is an easy-to-use assay based on highly specific monoclonal antibodies with insignificant or no cross-reactivity to C-peptide or proinsulin.
Catalog No. NC0067944