Learn More
Enzo Life Sciences Insulin receptor monoclonal antibody (9H4) (100 µg)
SDP
Supplier:  Enzo Life Sciences ADI905647100
Encompass_Preferred
Clone: 9H4. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of insulin receptor. UniProt: P06213. Species Reactivity: Human, Mouse, Rat, Dog. Applications: ELISA, IHC, WB. Recommended dilutions: ELISA (0.1µg/ml). Immunohistochemistry (1:50). Western Blot (0.1µg/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-658-403