Learn More
Eurofins Discoverx Insulin Receptor, active, 250 µg. (14-466M)
Supplier:  Eurofins Discoverx 14466M
Encompass
N-terminal 6xHis-tagged recombinant, human Insulin receptor amino acids 1005-1310. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8836