Learn More
Cell Signaling Technology Insulin Receptor Substrate Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502051180
Encompass
The Insulin Receptor Substrate Antibody Sampler Kit provides an economical solution for investigating IRS-1 and IRS-2 signaling and phosphorylation pathways within the cell. This kit is optimized for western blot analysis, enabling the simultaneous study of multiple phosphorylation sites and total protein levels.
  • Includes primary antibodies for detecting IRS-1 and IRS-2, as well as specific phospho-IRS-1 sites (Ser307, Ser612, and Ser318).
  • Contains secondary HRP-linked antibody for direct western blot detection.
  • Validated for use in western blotting applications.
  • Sufficient antibody quantity provided for two western blots per primary antibody.
  • Designed for research use in the study of insulin signaling, metabolism, and related pathways.
Catalog No. 50-205-1180 Supplier Cell Signaling Technology Supplier No. 3015T
The Insulin Receptor Substrate Antibody Sampler Kit provides an economical solution for investigating IRS-1 and IRS-2 signaling and phosphorylation pathways within the cell. This kit is optimized for western blot analysis, enabling the simultaneous study of multiple phosphorylation sites and total protein levels.
  • Includes primary antibodies for detecting IRS-1 and IRS-2, as well as specific phospho-IRS-1 sites (Ser307, Ser612, and Ser318).
  • Contains secondary HRP-linked antibody for direct western blot detection.
  • Validated for use in western blotting applications.
  • Sufficient antibody quantity provided for two western blots per primary antibody.
  • Designed for research use in the study of insulin signaling, metabolism, and related pathways.