Learn More
Crescent Chemical Co Inc INSULIN RECOMBINANT HUMAN
SDP
Supplier:  Crescent Chemical Co Inc SE26360.01
Encompass_Preferred
Insulin, recombinant human min. 27.5 USP U/mg Ph. Eur., USP. Identical in structure and function to the native human sequence. Essential for long-term growth of various cell lines. Stimulates the proliferation of cells and supports carbohydrate metabolism. Absence of insuli
Catalog No. 50-247-747