Learn More
Cayman Chemical Insulin Sensitizer C59, Purity: ≥98%, CAS: 2761446-81-3, 5 mg
SDP
Catalog No. 502945804
Encompass_Preferred
A potentiator of insulin-induced Glut4 plasma membrane translocation; enhances insulin-stimulated increases in plasma membrane levels of Glut4 in a reporter assay using C2C12 cells at 10 µM; decreases fasting blood levels of glucose, fasting plasma levels of insulin, and body weight in mice fed a high-fat diet at 10 mg/kg per day; potentiates insulin-induced increases in the plasma membrane translocation of Glut4 in the forelimbs, hind limbs, brown adipose tissues, and heart but not brain or liver in mice in a model of diet-induced obesity
Catalog No. 50-294-5804 Supplier Cayman Chemical Supplier No. 421295 MG
A potentiator of insulin-induced Glut4 plasma membrane translocation; enhances insulin-stimulated increases in plasma membrane levels of Glut4 in a reporter assay using C2C12 cells at 10 µM; decreases fasting blood levels of glucose, fasting plasma levels of insulin, and body weight in mice fed a high-fat diet at 10 mg/kg per day; potentiates insulin-induced increases in the plasma membrane translocation of Glut4 in the forelimbs, hind limbs, brown adipose tissues, and heart but not brain or liver in mice in a model of diet-induced obesity