Learn More
Sigma Aldrich Fine Chemicals Biosciences Insulin solution human, 11061-68-0, MFCD00131380, 5mL
Catalog No. 501656853
Encompass
Chemically defined, recombinant, expressed in Saccharomyces cerevisiae, sterile-filtered, BioXtra, suitable for cell culture
Catalog No. 50-165-6853 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I92785ML
Chemically defined, recombinant, expressed in Saccharomyces cerevisiae, sterile-filtered, BioXtra, suitable for cell culture