Learn More
Med Vet International Easy Touch U-100 Insulin Syringe, 1cc 29g x 1/2in, 100/Box
Catalog No. NC1551333
Encompass_Preferred
The Easy Touch U-100 Insulin syringe is designed and manufactured to deliver the highest level of comfort. Their special needles are triple-bevel cut to a super sharp point, electro-polished to remove microscopic burrs, and film-coated for smoother injection. The attached 29g 1/2" needle is color-coded to the gauge and the thin wall cannulas provide an easier flow. Color code: Green Syringe size: 1cc/1mL Needle gauge: 29g Needle size: 1/2in.
This item is not returnable. View return policy
Catalog No. NC1551333 Supplier Med Vet International Supplier No. 829155
The Easy Touch U-100 Insulin syringe is designed and manufactured to deliver the highest level of comfort. Their special needles are triple-bevel cut to a super sharp point, electro-polished to remove microscopic burrs, and film-coated for smoother injection. The attached 29g 1/2" needle is color-coded to the gauge and the thin wall cannulas provide an easier flow. Color code: Green Syringe size: 1cc/1mL Needle gauge: 29g Needle size: 1/2in.