Learn More
Enterprise Technology Solutions Insulin Syringe - Non-Toxic 30
SDP
Catalog No. 502886054
Encompass_Preferred
Insulin Syringe - Non-Toxic 30G 5/16 Needle 0.5 cc 5/100/cs
Catalog No. 50-288-6054 Supplier Enterprise Technology Solutions Supplier No. ETS6953
Insulin Syringe - Non-Toxic 30G 5/16 Needle 0.5 cc 5/100/cs