Learn More
Supply Solutions Insulin-Transferrin-Sodium Selenite Supplement, pkg of 50 mg (for 5 l medium)
Supplier:  Supply Solutions SIGMA 11074547001
Encompass
The Insulin-Transferrin-Sodium Selenite Supplement contains the most essential growth-promoting components of serum-free culture media in an optimized ratio.
Catalog No. 50-100-4927