Learn More
Medchemexpress LLC Insulin-Transferrin-Selenium, 100X | 10 ML
SDP
Supplier:  Medchemexpress LLC HY15028710ML
Encompass_Preferred
Insulin-Transferrin-Selenium, 100X (ITS-G) is a mixture of insulin, transferrin, and sodium selenite. It is a general supplement designed for use in classic media such as DMEM, RPMI-1640, and nutrient media like Ham's F-12, DMEM/F12. This supplement is added to basal medium to reduce the amount of fetal calf serum required to culture cells.
  • Designed for use in classic media such as DMEM, RPMI-1640, and nutrient media such as Ham's F-12, DMEM/F12
  • Reduces the amount of fetal calf serum required to culture cells
Catalog No. 50-003-75477