Learn More
Medchemexpress LLC Insulin-Transferrin-Selenium-Ethanolamine (ITS-X) | 10 ML
SDP
Supplier:  Medchemexpress LLC HY150287A10ML
Encompass_Preferred
Insulin-Transferrin-Selenium-Ethanolamine (ITS-X) is a basal medium supplement designed to reduce the amount of fetal bovine serum (FBS) required for cell culture. It is intended for research use only.
  • Basal medium supplement
  • Reduces fetal bovine serum (FBS) in cell culture
  • Liquid appearance
  • Colorless to light yellow
  • For research use only
Catalog No. 50-003-75571