Learn More
BioVendor, LLC Insulin, Ultrasensitive (0.025-1.5 ng/mL) mouse ELISA, 96 wells
SDP
Supplier:  BioVendor, LLC RAI007R
Encompass
Insulin, Ultrasensitive (0.025 1.5 ng/ml) mouse ELISA, 96 wells. Immunoassay kit for quantitative measurement of mouse insulin in serum, plasma, and body fluid samples. Calibration range: 0.2 - 7 ng/mL. Detection limit: 0.025 ng/ml. Summary: Insulin is a peptide hormone secreted from B cells of islet of Langerhans in the pancreas. Main targets of insulin are liver, muscle, and adipose tissue. In the liver, it promotes glycogenesis, protein synthesis, fatty acid synthesis, carbohydrate utilization, and inhibition of gluconeogenesis. In the muscle, it promotes membrane permeability for carbohydrates, amino acids and K ion, glycogenesis, protein synthesis, while inhibits protein degradation. In the adipose tissue, it promotes membrane permeability for glucose and fatty acid synthesis.
Catalog No. 50-206-9171