Learn More
Alpco Ltd Ultrasensitive Insulin ELISA
SDP
Supplier:  Alpco Ltd 80INSHUUE01.1
Encompass_Preferred
The Ultrasensitive Insulin ELISA quantifies the concentration of insulin protein in human serum or plasma samples. FDA Class I Device. For In Vitro Diagnostic Use.
  • Only 25 µL of sample is sufficient for the assay
  • Superior sensitivity of 0.135 µIU/mL
  • Dynamic range of 0.15-20 µIU/mL
Catalog No. 50-751-3655