Learn More
Cayman Chemical InsulInase InhIbItr 500uL
SDP
Catalog No. 502965043
Encompass_Preferred
A mAChR antagonist (Kis 0.42 1.77 0.52 0.78 and 1.29 nM for the M1-M5 receptors respectively) induces relaxation of precontracted isolated human bronchi at 0.01 0.1 or 1 UM reduces post-prandial gastric antral motility in dogs at 0.01 mg/kg inhibits salivation in a rat model of pilocarpine-induced sialorrhea (ED50 0.74 Ug/kg)
Catalog No. 50-296-5043 Supplier Cayman Chemical Supplier No. 18429500 UL
Add to Cart
Add to Cart
A mAChR antagonist (Kis 0.42 1.77 0.52 0.78 and 1.29 nM for the M1-M5 receptors respectively) induces relaxation of precontracted isolated human bronchi at 0.01 0.1 or 1 UM reduces post-prandial gastric antral motility in dogs at 0.01 mg/kg inhibits salivation in a rat model of pilocarpine-induced sialorrhea (ED50 0.74 Ug/kg)