Learn More
Supply Solutions Insulin, human, 100mg
Supplier:  Supply Solutions SIGMA 11376497001
Encompass
Recombinant Insulin, human, is produced in yeast and purified by standard chromatographic techniques. Insulin regulates blood glucose. In muscles and adipocytes it stimulates glucose intake and metabolism. It also controls the expression and various enzymes.
Catalog No. 50-100-3415