Learn More
ACROBiosystems Insulin Receptor Protein, Human, His-Tagged, 1 mg
Catalog No. 501957000
Encompass_Preferred
Human Insulin receptor (INSR), also known as CD220, is a disulfide-linked heterotetramer consisting of two alpha and two beta chains. This protein is expressed in human HEK293 cells and contains amino acids His 28 to Lys 944. It is designed for applications requiring high purity and low endotoxin levels.
  • Expressed from human HEK293 cells for authentic post-translational modifications
  • Verified by MALS and SPR for quality assurance
  • Low endotoxin level of less than 0.01 EU per μg
  • Purity ≥90% by SDS-PAGE and ≥95% by SEC-MALS
  • Lyophilized from a 0.22 μm filtered PBS solution, pH 7.4, with trehalose protectant
  • Carries a polyhistidine tag at the C-terminus
Catalog No. 50-195-7000 Supplier ACROBiosystems Supplier No. INRH52HA1MG
Human Insulin receptor (INSR), also known as CD220, is a disulfide-linked heterotetramer consisting of two alpha and two beta chains. This protein is expressed in human HEK293 cells and contains amino acids His 28 to Lys 944. It is designed for applications requiring high purity and low endotoxin levels.
  • Expressed from human HEK293 cells for authentic post-translational modifications
  • Verified by MALS and SPR for quality assurance
  • Low endotoxin level of less than 0.01 EU per μg
  • Purity ≥90% by SDS-PAGE and ≥95% by SEC-MALS
  • Lyophilized from a 0.22 μm filtered PBS solution, pH 7.4, with trehalose protectant
  • Carries a polyhistidine tag at the C-terminus