Learn More
Grainger InsulMultitapCon PVC 14 AWG 4 AWG
Catalog No. 502748813
Encompass_Preferred
The Polaris Insulated Multitap Connector is a 4-way insulated tap connector that allows multiple wires to be connected to a single wire. It is rated for 600 volts and can be used for a variety of residential and commercial applications including lighting, motors, appliances, and more. The connector body and cover are made of UV resistant polypropylene for durability. It has a temperature rating of 105°C and comes with a clear insulating cover to prevent accidental contact. This 4-way multitap connector helps simplify wiring and reduces installation time.
Catalog No. 50-274-8813 Supplier Grainger Supplier No. 14V971
The Polaris Insulated Multitap Connector is a 4-way insulated tap connector that allows multiple wires to be connected to a single wire. It is rated for 600 volts and can be used for a variety of residential and commercial applications including lighting, motors, appliances, and more. The connector body and cover are made of UV resistant polypropylene for durability. It has a temperature rating of 105°C and comes with a clear insulating cover to prevent accidental contact. This 4-way multitap connector helps simplify wiring and reduces installation time.