Learn More
Grainger InsulMultitapCon PVC 250 kcmil 750 kcmil
Catalog No. 502750642
Encompass_Preferred
The Polaris Insulated Multitap Connector is an electrical connector that allows multiple wires to be connected to a single point. It is rated for 600 volts and can accommodate wire sizes from 12 to 6 AWG. The connector has 4 ports and is made from thermoplastic polyester with nickel-plated brass terminals. It is UL listed and CSA certified. The multitap is insulated to help prevent shorts and is ideal for splicing and extending multiple branch circuits. This connector can be installed using hand tools for efficient wiring jobs.
Catalog No. 50-275-0642 Supplier Grainger Supplier No. 14V966
The Polaris Insulated Multitap Connector is an electrical connector that allows multiple wires to be connected to a single point. It is rated for 600 volts and can accommodate wire sizes from 12 to 6 AWG. The connector has 4 ports and is made from thermoplastic polyester with nickel-plated brass terminals. It is UL listed and CSA certified. The multitap is insulated to help prevent shorts and is ideal for splicing and extending multiple branch circuits. This connector can be installed using hand tools for efficient wiring jobs.