Learn More
Grainger InsulMultitapCon PVC 4 AWG 500 kcmil
Catalog No. 502750989
Encompass_Preferred
The Polaris Insulated Multitap Connector is a 14-gauge, 3-wire multitap connector with a maximum voltage of 600V and maximum amperage of 15A per wire. It has a maximum conductor range of #14-10 AWG and is made of thermoplastic polyvinyl chloride (PVC) with a temperature rating of 105°C. This UL-listed connector has 3 ports and is designed for splicing and extending 3-wire jacketed cables. It measures 2.56" x 1.5" x 1.25" and features a clear insulating cover for safety. This multitap connector provides a reliable electrical connection for extending and splicing cables.
Catalog No. 50-275-0989 Supplier Grainger Supplier No. 14V982
The Polaris Insulated Multitap Connector is a 14-gauge, 3-wire multitap connector with a maximum voltage of 600V and maximum amperage of 15A per wire. It has a maximum conductor range of #14-10 AWG and is made of thermoplastic polyvinyl chloride (PVC) with a temperature rating of 105°C. This UL-listed connector has 3 ports and is designed for splicing and extending 3-wire jacketed cables. It measures 2.56" x 1.5" x 1.25" and features a clear insulating cover for safety. This multitap connector provides a reliable electrical connection for extending and splicing cables.