Learn More
Grainger InsulMultitapCon PVC 4 AWG 500 kcmil
Catalog No. 502750965
Encompass_Preferred
The Polaris Insulated Multitap Connector is a heavy duty electrical connector that allows multiple wires to be connected to a single point. It is made of high impact nylon for durability and has an IP68 waterproof rating. The connector has 4 ports that accept wire sizes from 16-6 AWG and is rated for 600V and 30A. It can be used for a variety of automotive, marine, and industrial applications. The unique seal design keeps moisture out. This product is UL listed and made in the USA.
Catalog No. 50-275-0965 Supplier Grainger Supplier No. 14V981
The Polaris Insulated Multitap Connector is a heavy duty electrical connector that allows multiple wires to be connected to a single point. It is made of high impact nylon for durability and has an IP68 waterproof rating. The connector has 4 ports that accept wire sizes from 16-6 AWG and is rated for 600V and 30A. It can be used for a variety of automotive, marine, and industrial applications. The unique seal design keeps moisture out. This product is UL listed and made in the USA.