Learn More
Grainger InsulMultitapCon PVC 6 AWG 250 kcmil
Catalog No. 502750971
Encompass_Preferred
The Polaris insulated multitap wire connectors are designed for connecting and splicing multiple wires. They feature a split bolt design that allows wires to be inserted into the connector without disassembling it. The connectors have an insulated housing that helps prevent shorts and are UL listed. They come in a variety of sizes to accommodate different wire gauges and number of wires. The split bolt design allows for a secure mechanical connection and the insulation helps prevent corrosion. Overall, these connectors provide a safe and reliable method for splicing multiple wires in an electrical system.
Catalog No. 50-275-0971 Supplier Grainger Supplier No. 14V977
The Polaris insulated multitap wire connectors are designed for connecting and splicing multiple wires. They feature a split bolt design that allows wires to be inserted into the connector without disassembling it. The connectors have an insulated housing that helps prevent shorts and are UL listed. They come in a variety of sizes to accommodate different wire gauges and number of wires. The split bolt design allows for a secure mechanical connection and the insulation helps prevent corrosion. Overall, these connectors provide a safe and reliable method for splicing multiple wires in an electrical system.