Learn More
Grainger InsulMultitapCon PVC 6 AWG 250 kcmil
Catalog No. 502749538
Encompass_Preferred
The insulated PVC multitap mechanical split bolt connectors from Grainger are designed for splicing and tapping insulated wires. They feature a tapered split bolt design that tightens around conductors when the nut is tightened. The insulation sleeve helps prevent shorts and corrosion. These connectors are UL listed for copper and aluminum conductors sized 14-6 AWG and are rated up to 600 volts. They come in a variety of tap configurations from 2-way up to 12-way. The PVC insulation provides good chemical and moisture resistance. Overall, these connectors offer a secure and insulated method for splicing multiple wires in industrial and commercial applications.
Catalog No. 50-274-9538 Supplier Grainger Supplier No. 14V975
The insulated PVC multitap mechanical split bolt connectors from Grainger are designed for splicing and tapping insulated wires. They feature a tapered split bolt design that tightens around conductors when the nut is tightened. The insulation sleeve helps prevent shorts and corrosion. These connectors are UL listed for copper and aluminum conductors sized 14-6 AWG and are rated up to 600 volts. They come in a variety of tap configurations from 2-way up to 12-way. The PVC insulation provides good chemical and moisture resistance. Overall, these connectors offer a secure and insulated method for splicing multiple wires in industrial and commercial applications.