Learn More
Grainger InsulMultitapCon PVC 6 AWG 3/0 AWG
Catalog No. 502749561
Encompass_Preferred
The Polaris Insulated Multitap Connector is an electrical connector that allows multiple wires to be connected to a single point. It is rated for 600 volts and can accommodate wire sizes from 14-10 AWG. The connector has a maximum temperature rating of 105°C and is made from high-quality thermoplastic. It features a lever-style wire clamp for easy installation and secure wire retention. The connector body measures 2.56" x 1.5" x 1.31" and has 4 ports to connect multiple wires. It can be mounted via the .31" diameter hole on the connector body. The connector meets UL and CSA standards for safety.
Catalog No. 50-274-9561 Supplier Grainger Supplier No. 14V973
The Polaris Insulated Multitap Connector is an electrical connector that allows multiple wires to be connected to a single point. It is rated for 600 volts and can accommodate wire sizes from 14-10 AWG. The connector has a maximum temperature rating of 105°C and is made from high-quality thermoplastic. It features a lever-style wire clamp for easy installation and secure wire retention. The connector body measures 2.56" x 1.5" x 1.31" and has 4 ports to connect multiple wires. It can be mounted via the .31" diameter hole on the connector body. The connector meets UL and CSA standards for safety.