Learn More
Grainger InsulMultitapCon PVC 6 AWG 350 kcmil
Catalog No. 502750986
Encompass_Preferred
The Polaris Insulated Multitap Wire Connectors are designed for connecting multiple wires in electrical applications. They feature a split bolt design that allows wires to be inserted into the connector and tightened for a secure connection. The insulated housing helps prevent shorts and shocks. They are UL listed for safety and are available in a range of sizes to accommodate different wire gauges and number of wires. The connectors have a maximum temperature rating of 105°C and maximum voltage rating of 600V. They are constructed from plated steel with a thermoplastic housing.
Catalog No. 50-275-0986 Supplier Grainger Supplier No. 14V978
The Polaris Insulated Multitap Wire Connectors are designed for connecting multiple wires in electrical applications. They feature a split bolt design that allows wires to be inserted into the connector and tightened for a secure connection. The insulated housing helps prevent shorts and shocks. They are UL listed for safety and are available in a range of sizes to accommodate different wire gauges and number of wires. The connectors have a maximum temperature rating of 105°C and maximum voltage rating of 600V. They are constructed from plated steel with a thermoplastic housing.