Learn More
Grainger InsulMultitapCon PVC 6 AWG 350 kcmil
Catalog No. 502750988
Encompass_Preferred
The Polaris Insulated Multitap Connector is an electrical connector that allows multiple wires to be connected to a single source. It is rated for 600 volts and can accommodate wire sizes from 12 to 6 AWG. The connector has 4 ports and is made from UV resistant polycarbonate. It can be used for a variety of indoor and outdoor applications including landscape lighting, pools, spas, and more. The connector measures 4.5 x 2.75 x 1.5 inches and has an operating temperature range of -40°F to 221°F. It is cULus listed for safety.
Catalog No. 50-275-0988 Supplier Grainger Supplier No. 14V980
The Polaris Insulated Multitap Connector is an electrical connector that allows multiple wires to be connected to a single source. It is rated for 600 volts and can accommodate wire sizes from 12 to 6 AWG. The connector has 4 ports and is made from UV resistant polycarbonate. It can be used for a variety of indoor and outdoor applications including landscape lighting, pools, spas, and more. The connector measures 4.5 x 2.75 x 1.5 inches and has an operating temperature range of -40°F to 221°F. It is cULus listed for safety.